3-Amino-5-(trifluoromethyl)benzonitrile ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A193579-5G
A193579-1G
A193579-250MG
ALA193579-5GEA
290.61
USD
ALA193579-5G
ALA193579-1G
ALA193579250MG
3-Amino-5-(trifluoromethyl)benzonitrile ≥98%
3-Amino-5-(trifluoromethyl)benzonitrile
Formula:
C8H5F3N2 MW: 186.1339 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09955455 CAS Number: 49674-28-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...