3-Amino-3-(3'-pyridyl)propionic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Amino-3-(3-pyridyl)propionic Acid, (RS)-3-Amino-3-(3-pyridyl)-propionic acid

A151148-100G A151148-1G A151148-25G
ALA151148-100GEA 3750.51 USD
ALA151148-100G ALA151148-1G ALA151148-25G
3-Amino-3-(3'-pyridyl)propionic acid ≥98% (by HPLC)
3-Amino-3-(3'-pyridyl)propionic acid
Formula: C8H10N2O2
Melting Pt: 207 °C
MDL Number: MFCD01549598
CAS Number: 62247-21-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83c44e4a93c3ee89ccb1042e16e362818bd0bb09&vid=83c44e4a93c3ee89ccb1042e16e362818bd0bb09&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR