1-Fluoro-2-methoxy-4-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

90191-5G 90191-25G 90191-1G 90191-500MG
77633-830EA 792 USD
77633-830 77660-514 77644-184 77652-184
1-Fluoro-2-methoxy-4-(methoxymethoxy)benzene ≥95%
1-Fluoro-2-methoxy-4-(methoxymethoxy)benzene

Caution: For Research Use Only

Formula: C₉H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34772156
CAS Number: 2120398-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR