Conductivity Standards
Supplier: Hach
210542 2307442 2307542 1440049 1440026 1440042 LZW9700.99 LZW9710.99 LZW9720.99
104085-766EA 43.92 USD
104085-766 104086-092 104086-094 104085-462 104085-458 104085-460 104088-628 104088-632 104088-636
Conductivity Standards
Standards Conductivity Standards
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.