Sodium periodate

Supplier: Hach
Danger

Synonyms: Sodium metaperiodate

98499
104088-380PK 66.58 USD
104088-380
Sodium periodate
Sodium periodate
Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 175 °C
Density: 3.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1,II
Merck Index: 13,08714

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR