Phenolphthalein 0.5% in aqueous solution

Supplier: Hach

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

16253
104085-594EA 94.81 USD
104085-594
Phenolphthalein 0.5% in aqueous solution
Phenolphthalein
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
https://us.vwr.com/stibo/web/std.lang.all/72/01/2297201.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR