Methyl orange 0.5 g/L in aqueous solution
Supplier: Hach
Synonyms:
4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt
, Methyl orange sodium salt
, Helianthine
, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt
14849
104085-570EA
50.86
USD
104085-570
Methyl orange 0.5 g/L in aqueous solution
Methyl orange
Formula:
C₁₄H₁₄N₃NaO₃S MW: 327.34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007502 CAS Number: 547-58-0 EINECS: 208-925-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...