Citric acid
Supplier: Hach
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid
, Hydroxytricarballylic acid
1454828
104085-498PK
658.32
USD
104085-498
Citric acid
Citric acid
Formula:
HOC(COOH)(CH₂COOH)₂ MW: 192.13 g/mol Boiling Pt: 310 °C (1013 hPa) Melting Pt: 149…151 °C Density: 1.665 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 201-069-1 REACH: 01-2119457026-42-0000 Merck Index: 13,02350 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...