2,3-Dichloro-6-fluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

37580-5G 37580-25G
77634-946EA 148 USD
77634-946 77655-484
2,3-Dichloro-6-fluorobenzonitrile ≥97%
2,3-Dichloro-6-fluorobenzonitrile

Caution: For Research Use Only

Formula: C₇H₂Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD04115878
CAS Number: 79544-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR