1-Bromo-2-((3-(tert-butyl)phenoxy)methyl)benzene ≥97%

Supplier: AOBChem USA

Warning

335315-5G 335315-1G 335315-25G 335315-500MG
77628-664EA 1707.74 USD
77628-664 77638-170 77655-152 77646-150
1-Bromo-2-((3-(tert-butyl)phenoxy)methyl)benzene ≥97%
1-Bromo-2-((3-(tert-butyl)phenoxy)methyl)benzene

Caution: For Research Use Only

Formula: C₁₇H₁₉BrO
Storage Temperature: Ambient
MDL Number: MFCD18918328
CAS Number: 877663-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR