2-Fluoro-3-iodo-6-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

223735-5G 223735-25G 223735-1G 223735-500MG
77628-140EA 1484.23 USD
77628-140 77658-050 77641-286 77649-324
2-Fluoro-3-iodo-6-(methylthio)pyridine ≥95%
2-Fluoro-3-iodo-6-(methylthio)pyridine

Caution: For Research Use Only

Formula: C₆H₅FINS
Storage Temperature: Ambient
CAS Number: 1803851-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR