RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: 29F.1A12™-CP159]

Supplier: Bio X Cell
CP159-100MG CP159-1MG CP159-25MG CP159-50MG CP159-5MG
77580-340EA 6269.61 USD
77580-340 77580-342 77580-344 77580-346 77580-348
RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: 29F.1A12™-CP159]
Antibodies
Type: Primary
Antigen: PD-1
Clonality: Recombinant
Clone: 29F.1A12™-CP159
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR