InVivoSIM Anti-amyloid-beta Human Antibody

Supplier: Bio X Cell
SIM0032-100MG SIM0032-1MG SIM0032-25MG SIM0032-50MG SIM0032-5MG
77580-820EA 7984.56 USD
77580-820 77580-822 77580-824 77580-826 77580-828
InVivoSIM Anti-amyloid-beta Human Antibody
Antibodies
Lecanemab biosimilar.

Type:
Antigen: amyloid-beta
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR