2-Fluorophenethylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(2-Fluorophenyl)ethylamine2-Fluorophenethylamine1-Amino-2-(2-fluorophenyl)ethane

350930050
TS350930050EA 37.4 USD
TS350930050
2-Fluorophenethylamine 97%
2-Fluorophenethylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 64 °C (.6 mmHg)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00075502
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow to pink-orange Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR