N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetic acid dimethylamide
, Dimethyl Acetamide
, DMAC
390800010
TS390800010EA
104
USD
TS390800010
N,N-Dimethylacetamide 99.5+% for analysis
N,N-Dimethylacetamide
Formula:
CH₃CON(CH₃)₂ MW: 87.12 g/mol Boiling Pt: 164…166 °C (760 mmHg) Melting Pt: –20 °C Density: 0.937 g/cm³ Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008686 CAS Number: 127-19-5 EINECS: 204-826-4 |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.05 % |
GC | ≥99.50 % |
Residue after evaporation | ≤0.0005 % |
Acidity | ≤0.0005 meq/g |
Chloride (Cl) | ≤0.001 % |
Sulfate (SO4) | ≤0.001 % |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...