Dichloromethane 99.8% stabilized for HPLC

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methane dichlorideMethylene chloride

Stabilization: Stabilised with amylene

268330025 268330010
TS268330010EA 78.1 USD
TS268330010 TS268330025
Dichloromethane 99.8% stabilized for HPLC
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –97 °C
Density: 1.325 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.02 %
GC ≥99.8 %
UV at 233 nm A: ≤1
UV at 240 nm A: ≤0.2
UV at 250 nm A: ≤0.02
UV at 260 nm A: ≤0.005
UV at 270 nm A: ≤0.005
UV at 280 nm A: ≤0.005
UV 1 cm cell vs HPLC-grade water
Color scale ≤10 APHA
Stabilizer About 100 ppm Amylene
Residue after evaporation ≤0.0002 %
Acidity ≤0.0001 meq/g
Filtered to 0.2 micrometer Filtered
Free chlorine ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR