5-Bromoindane ≥95%

Supplier: Ambeed
Danger

A924883-1G A924883-250MG
77573-850EA 110.21 USD
77573-850 77674-376
5-Bromoindane ≥95%
5-Bromoindane

Caution: for Research Use Only

Formula: C₉H₉Br
MDL Number: MFCD09842508
CAS Number: 6134-54-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR