Coelenterazine 400a ≥97%

Supplier: Ambeed
Warning

Synonyms: BlueSyn C2,8-Dibenzyl-6-phenylimidazo[1,2-a]pyrazin-3(7H)-one2,8-dibenzyl-6-phenyl-7H-imidazo[1,2-a]pyrazin-3-oneDeepBlue C™Coelenteramine 400a

A1210625-5MG A1210625-10MG A1210625-1MG
77596-324EA 279.83 USD
77596-324 77596-326 77596-328
Coelenterazine 400a ≥97%
Coelenterazine 400a

Caution: for Research Use Only

Formula: C₂₆H₂₁N₃O
MW: 391.46 g/mol
MDL Number: MFCD04114093
CAS Number: 70217-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR