10-Nonadecanol ≥95%

Supplier: Ambeed
Warning

Synonyms: Nonadecan-10-ol10-Nonadecyl alcohol10-Hydroxy nonadecane

A201140-1G
77550-288EA 135.96 USD
77550-288
10-Nonadecanol ≥95%
10-Nonadecanol

Caution: for Research Use Only

Formula: C₁₉H₄₀O
MW: 284.53 g/mol
CAS Number: 16840-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR