Western Transfer Protein System, IBI Scientific
Supplier: IBI Scientific
IB95000
14225-088EA 875.17 USD
14225-088
Western Transfer Protein System, IBI Scientific
Blotting Apparatus
Use this system for rapid transfer of protein/peptide samples or nucleic acids from mini polyacrylamide gels to 8x10cm (33/16x315/16") or 10x10cm (315/16x315/16") membranes
The unit accommodates two cassette sandwiches for transfer. Proteins from mini PAGE gels can be transferred within the electrode module in 60 to 90 minutes. The unit consists of two hinged perforated transfer cassettes, each color-coded with 3.2mm (1/8”) sponges and a lock. The cassettes are constructed of molded ABS and slide into an electrode module with a distance of 4.5cm (13/4") between the electrodes. The blotting application is dependent on the voltage applied over the electrodes. Therefore, the distance between the electrodes is kept at a minimum for a more efficient protein transfer. (IBI IB95000)