4-Formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Formylphenyl)metaboric acid4-Formylbenzeneboronic acid4-BoronobenzaldehydeDihydroxy(4-formylphenyl)borane

10561-25G
77497-574EA 64.38 USD
77497-574
4-Formylphenylboronic acid ≥97%
4-Formylphenylboronic acid

Caution: for Research Use Only

Formula: C₇H₇BO₃
MW: 149.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151823
CAS Number: 87199-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR