VWR® TraceClean® Containers with Chemical Preservative
Supplier: VWR
VWR® TraceClean®
QLPC89080072 QLPW89080074 QLFS89080080
89094-186CS 101.91 USD
89094-186 89094-188 89094-200
VWR® TraceClean® Containers with Chemical Preservative
Bottles Water Sampling Bottles
Containers are filled with chemical preservative to maintain sample integrity until analysis can be performed.
  • Containers are made of HDPE or glass
  • Provided with polypropylene cap
  • Cap may or may not have a liner
  • Hydrochloric acid and sulfuric acid are for oil
HDPE containers are for water samples. Bottle 89094-186 and packer 89094-188 are supplied with polyfoam-lined polypropylene cap, and NALGENE® jars are supplied with linerless polypropylene cap. Glass jars are supplied with polypropylene cap with 3.2 mm (0.125") fluoropolymer liner.
All containers are Quality-Assured® (QA) and follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. Containers include a Certificate of Analysis in every case. Containers also include bar codes with lot number and individual container number for complete traceability.
All cases are custody-sealed to ensure content integrity. Alternate preservative volumes may be substituted.