Iron, annular sputtering target, Diameter 59×33 mm (OD×ID)

Supplier: Electron Microscopy Sciences

91032-FE
102099-854EA 568.02 USD
102099-854
Iron, annular sputtering target, Diameter 59×33 mm (OD×ID)
Iron

Metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.

Formula: Fe
MW: 55.85 g/mol
MDL Number: MFCD00010999
CAS Number: 7439-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR