N,N-Dimethylacetamide, anhydrous ≥99.9%, DriSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid dimethylamideDimethyl AcetamideDMAC

Packed in septum-sealed bottle.

DX1544-6 DX1544-6
EM-DX1544-6CS 2193.9 USD
EM-DX1544-6 CADX1544-6
N,N-Dimethylacetamide, anhydrous ≥99.9%, DriSolv®, Supelco®
N,N-Dimethylacetamide

Clear and free from particulates. Suitable for use in organic synthesis and other anhydrous solvent applications.
Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2) for use with elbow adapter minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.

Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 166.1 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03254
https://us.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg

Order Now

Specification Test Results

Assay (GC) 99.8% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Residue after evaporation 10ppm max
Water (H2O) 50ppm max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR