Potassium acetate ≥99.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
Synonyms:
Diuretic salt
, 4-Chloro-N-furfuryl-5-sulfamoylanthranlic acid
, Acetic acid potassium salt
PX1330-30
PX1330-7
PX1330-1
PX1330-3
PX1330-3
PX1330-1
PX1330-30
EM-PX1330-30EA
3309.42
USD
EM-PX1330-30
CAPX1330-3
CAPX1330-1
EM-PX1330-1
EM-PX1330-3
CAPX1330-30
EM-PX1330-7
Potassium acetate ≥99.0%, GR ACS, Supelco®
Potassium acetate
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Formula:
CH₃COOK MW: 98.14 g/mol Melting Pt: 304 °C Density: 1.57 g/cm³ (25 °C) Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012458 CAS Number: 127-08-2 EINECS: 204-822-2 Merck Index: 13,07687 |
Specification Test Results
Assay | 99.0% min |
Calcium (Ca) | 0.005% max |
Chloride (Cl) | 0.003% max |
Color | White |
Form | Crystals or crystalline powder |
Heavy metals (as Pb) | 5ppm max |
Insoluble matter | 0.005% max |
Iron (Fe) | 5ppm max |
Magnesium (Mg) | 0.002% max |
pH of a 5% solution at 25°C | 6.5-9.0 |
Phosphate (PO4) | 0.001% max |
Sodium (Na) | 0.03% max |
Sulfate (SO4) | 0.002% max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...