Iron(III) citrate monohydrate, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Iron (III) citrate tribasic monohydrate
, Ferric citrate monohydrate
, Ferric Citrate, n-Hydrate
Management of Change (MOC) category = N
BAKRM376-07
BAKRM376-07
JTM376-7CS
3388.94
USD
JTM376-7
CAJTM376-7
Iron(III) citrate monohydrate, J.T.Baker®
Iron (III) citrate monohydrate
Formula:
C₆H₅FeO₇·H₂O MW: 262.96 g/mol |
MDL Number:
MFCD00013098 CAS Number: 2338-05-8 EINECS: 249-117-0 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (as Fe) | ≥16.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...