Razor Blades, Accutec Blades
Supplier: AccuTec Blades
94-0451 62-0314 95-0044-0000 62-0097 62-0170-0000 62-0315 62-0317 95-0043-0000
80085-960CS 1618.05 USD
80085-960 55411-060 55411-045 89029-794 80085-954 89087-536 80087-045 89031-954
Razor Blades, Accutec Blades
Razors and Blades
All blades are industrial grade
Single-edge blades are 0.23mm (0.009") thick and feature an extra-keen edge. Blades are available with steel or aluminum spines. Double-edge blades are 0.1mm (0.004") thick and are oiled.
Ordering information: Blades 80085-954 come individually wrapped in boxes of 250 blades with 20 boxes per case. Blades 80085-960 and 80087-045 are provided in dispenser packs of 10. Blades 89029-794 are designed for cleanroom applications and provided in dispenser packs of 10 with special sealed packaging. Blades 89087-536 come in boxes of 225 blades with 25 boxes per case. Blades 95062-668 are provided in dispenser packs of 50.