4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: PP 3

OR2650T-100MG OR2650T-250MG OR2650T-25MG
77407-800EA 72.34 USD
77407-800 77407-802 77407-804
4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine, Technical Grade
4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine

Chemiluminescent with antifilarial & affinity for the adenosine receptor.

Formula: C₁₁H₉N₅
Melting Pt: 214…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00022901
CAS Number: 5334-30-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR