1,3-Benzodioxole-5,6-diamine, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

OR110848-100MG OR110848-1G OR110848-500MG
77403-508EA 92.72 USD
77403-508 77403-510 77403-512
1,3-Benzodioxole-5,6-diamine, Technical Grade
1,3-Benzodioxole-5,6-diamine
Formula: C₇H₈N₂O₂
Melting Pt: 95…97 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD07780137
CAS Number: 38608-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR