2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,2,2-Trifluoro-1-(2-(trifluoromethyl)phenyl)ethanone, 2,2,2-Trifluoro-1-[2-(trifluoromethyl)phenyl]ethan-1-one, 2-(Trifluoroacetyl)benzotrifluoride
65656-1G
65656-5G
65656-25G
77340-908EA
62.83
USD
77340-908
77368-642
77380-854
2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone ≥95%
2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone
Caution: for Research Use Only
Formula:
C₉H₄F₆O MW: 242.118 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07380778 CAS Number: 42452-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...