6-Chloro-5-nitropyridine-3-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

34720-500MG 34720-1G 34720-5G 34720-25G
77325-706EA 190.55 USD
77325-706 77349-390 77362-058 77385-078
6-Chloro-5-nitropyridine-3-boronic acid pinacol ester ≥95%
6-Chloro-5-nitropyridine-3-boronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₁H₁₄BClN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD08063081
CAS Number: 1310383-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR