Anti-CD4 Rabbit Polyclonal Antibody

Supplier: ProSci
13-153
76761-276EA 435.69 USD
76761-276
Anti-CD4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR