Anti-XmAb-13676 Recombinant Antibody

Supplier: ProSci
10-951
76760-154EA 514.7 USD
76760-154
Anti-XmAb-13676 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR