Anti-LY2127399 Recombinant Antibody

Supplier: ProSci
10-470
76749-276EA 514.7 USD
76749-276
Anti-LY2127399 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR