10-Nonadecanone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Caprinone
, Dinonyl ketone
A252831-1G
A252831-5G
77193-686EA
60.77
USD
77193-686
77193-684
10-Nonadecanone 95%
10-Nonadecanone
Caution: for Research Use Only
Formula:
C₁₉H₃₈O MW: 282.51 g/mol |
MDL Number:
MFCD00009584 CAS Number: 504-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...