GSK429286A 98%

Supplier: Ambeed
Danger

Synonyms: N-(6-Fluoro-1H-indazol-5-yl)-2-methyl-6-oxo-4-(4-(trifluoromethyl)phenyl)-1,4,5,6-tetrahydropyridine-3-carboxamide, 4-[4-(Trifluoromethyl)phenyl]-N-(6-Fluoro-1H-indazol-5-yl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxamide, GSK429286, RHO-15

A569516-100MG A569516-10MG A569516-1MG A569516-25MG A569516-50MG A569516-5MG
77103-674EA 336 USD
77103-674 77103-676 77103-678 77103-680 77103-682 77103-684
GSK429286A 98%
GSK429286A

Caution: for Research Use Only

Formula: C₂₁H₁₆F₄N₄O₂
MDL Number: MFCD16618404
CAS Number: 864082-47-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR