2-Bromo-4,5-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-4,5-difluorophenyl)methanol

27738-1G 27738-25G 27738-500MG 27738-5G
76900-776EA 140.04 USD
76900-776 76900-778 76900-780 76900-782
2-Bromo-4,5-difluorobenzyl alcohol ≥97%
2-Bromo-4,5-difluorobenzyl alcohol

Caution: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18909667
CAS Number: 476620-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR