2-Fluoro-5-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

14537-25G 14537-5G
76895-552EA 453.01 USD
76895-552 76895-554
2-Fluoro-5-hydroxybenzaldehyde ≥97%
2-Fluoro-5-hydroxybenzaldehyde

Caution: for Research Use Only

Formula: C₇H₅FO₂
MW: 140.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038469
CAS Number: 103438-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR