3-Bromo-2,6-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23912-25G 23912-5G
76911-410EA 233.4 USD
76911-410 76911-412
3-Bromo-2,6-difluorobenzoic acid ≥97%
3-Bromo-2,6-difluorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₃BrF₂O₂
MW: 237.0000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261248
CAS Number: 28314-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR