2-Bromo-3-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: [2-Bromo-3-(trifluoromethyl)phenyl]methanol, (2-Bromo-3-(trifluoromethyl)phenyl)methanol

28890-1G 28890-25G 28890-500MG 28890-5G
76886-592EA 144.29 USD
76886-592 76886-594 76886-596 76886-598
2-Bromo-3-(trifluoromethyl)benzyl alcohol ≥97%
2-Bromo-3-(trifluoromethyl)benzyl alcohol

Caution: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185396
CAS Number: 1214330-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR