2-Ethoxybenzamide 98%

Supplier: Ambeed
Warning

Synonyms: Ethenzamide

A658200-1G
76879-372EA 5.31 USD
76879-372
2-Ethoxybenzamide 98%
2-Ethoxybenzamide

Caution: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00007977
CAS Number: 938-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR