Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Bis(2-cyanoethyl) diisopropylphosphoramidite
, Bis(2-cyanoethoxy)(diisopropylamino)phosphine
, Bis(2-cyanoethoxy)-N,N-diisopropylaminophosphine
A429168-5G
A429168-25G
A429168-1G
77551-288EA
91
USD
77551-288
76829-518
77551-290
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite 98%
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite
Caution: for Research Use Only
Formula:
C₁₂H₂₂N₃O₂P MW: 271.301 g/mol |
MDL Number:
MFCD00797597 CAS Number: 102690-88-0 UN: 2810 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...