N-Benzylguanidine compound with sulfuric acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzylguanidine hemisulphate, 1-Benzylguanidine hemisulfate

A847012-1G A847012-250MG A847012-5G
76698-370EA 45 USD
76698-370 76698-372 76698-374
N-Benzylguanidine compound with sulfuric acid 97%
N-Benzylguanidine compound with sulfuric acid

Caution: for Research Use Only

Formula: C₁₆H₂₄N₆O₄S
MDL Number: MFCD01746693
CAS Number: 2551-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR