Formononetin

Supplier: MilliporeSigma

Synonyms: 7-Hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one7-Hydroxy-3-(4-methoxyphenyl)chromone7-Hydroxy-4'-methoxyisoflavone

344215-5MG 344215-5MG
80502-492EA 116.53 USD
80502-492 CA80502-492
Formononetin
Formononetin
Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
MDL Number: MFCD00016948
CAS Number: 485-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR