4-Amino-2,3-difluorobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Carboxy-2,3-difluoroaniline

PC9169-1G PC9169-5G
76610-176EA 419.21 USD
76610-176 76610-178 76610-180
4-Amino-2,3-difluorobenzoic acid
4-Amino-2,3-difluorobenzoic acid
Formula: C₇H₅F₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD04114097
CAS Number: 194804-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR