4-Chloro-2-fluoro-5-methoxyaniline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Amino-2-chloro-4-fluoroanisole

PC500247-1G PC500247-250MG PC500247-5G
76605-374EA 206.78 USD
76605-374 76604-626 76606-828
4-Chloro-2-fluoro-5-methoxyaniline
4-Chloro-2-fluoro-5-methoxyaniline
Formula: C₇H₇ClFNO
MW: 175.588 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17169196
CAS Number: 91167-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR