2-Chloro-4-fluoro-5-iodophenol

Supplier: APOLLO SCIENTIFIC
Warning

PC500243-1G PC500243-250MG PC500243-5G
76607-740EA 128.36 USD
76607-740 76604-380 76607-516
2-Chloro-4-fluoro-5-iodophenol
2-Chloro-4-fluoro-5-iodophenol
Formula: C₆H₃ClFIO
MW: 272.443 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628048
CAS Number: 148254-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR