4-Bromo-2-fluoro-5-nitroanisole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-5-fluoro-4-methoxy-2-nitrobenzene

PC500241-1G PC500241-250MG
76605-722EA 48.9 USD
76605-722 76604-922
4-Bromo-2-fluoro-5-nitroanisole
4-Bromo-2-fluoro-5-nitroanisole
Formula: C₇H₅BrFNO₃
MW: 250.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100974
CAS Number: 661463-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR