6-Bromo-2-hydroxy-3-nitrobenzotrifluoride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Bromo-6-nitro-2-(trifluoromethyl)phenol, 3-Bromo-6-nitro-α,α,α-trifluoro-o-cresol

PC49607-1G PC49607-250MG
76604-616EA 574.9 USD
76604-616 76604-736
6-Bromo-2-hydroxy-3-nitrobenzotrifluoride 97%
6-Bromo-2-hydroxy-3-nitrobenzotrifluoride
Formula: C₇H₃BrF₃NO₃
MW: 286.003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25541885
CAS Number: 1445995-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR