2-Bromo-4,5-difluorobenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2-Bromo-4,5-difluorophenyl)methanol

PC49498-1G PC49498-5G
76607-144EA 115.14 USD
76607-144 76606-370
2-Bromo-4,5-difluorobenzyl alcohol
2-Bromo-4,5-difluorobenzyl alcohol
Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Melting Pt: 75…76 °C
Storage Temperature: Ambient
MDL Number: MFCD18909667
CAS Number: 476620-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR